Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGAGACATTTATTCTTTACTCA[A/G]TCCATAACAAGGAGGTTTGGGTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300547 | ||||||||||||||||||||
Literature Links: |
STK26 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STK26 - serine/threonine protein kinase 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042452.1 | Intron | NP_001035917.1 | ||||
NM_001042453.1 | Intron | NP_001035918.1 | ||||
NM_016542.3 | Intron | NP_057626.2 | ||||
XM_011531349.1 | Intron | XP_011529651.1 | ||||
XM_011531350.1 | Intron | XP_011529652.1 |