Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTTTCTAGGAAAAATGGCATTC[C/T]ATTTTAAAAAGTCATCATAGTATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179502 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAP1GDS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAP1GDS1 - Rap1 GTPase-GDP dissociation stimulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100426.1 | Intron | NP_001093896.1 | ||||
NM_001100427.1 | Intron | NP_001093897.1 | ||||
NM_001100428.1 | Intron | NP_001093898.1 | ||||
NM_001100429.1 | Intron | NP_001093899.1 | ||||
NM_001100430.1 | Intron | NP_001093900.1 | ||||
NM_021159.4 | Intron | NP_066982.3 |