Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAACATTTCATTCTGAAAGAGGCA[G/T]TTATGAACTAGAAAAAAAATGCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164011 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105377621 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105377621 - uncharacterized LOC105377621 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFKB1 - nuclear factor kappa B subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165412.1 | Intron | NP_001158884.1 | ||||
NM_001319226.1 | Intron | NP_001306155.1 | ||||
NM_003998.3 | Intron | NP_003989.2 | ||||
XM_011532006.2 | Intron | XP_011530308.1 | ||||
XM_011532008.2 | Intron | XP_011530310.1 |