Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTTTGAAATGCTTTTACACAGC[A/G]TTACAATTATTTTGGCTTTAGACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601213 MIM: 613745 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ABCE1 - ATP binding cassette subfamily E member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040876.1 | Intron | NP_001035809.1 | ||||
NM_002940.2 | Intron | NP_002931.2 |
ANAPC10 - anaphase promoting complex subunit 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |