Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAATAAAGTTAGGAGTGATACAT[C/T]CTAGAAAAGCACCTAAATACTTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 139397 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GUCY1B3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GUCY1B3 - guanylate cyclase 1 soluble subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000857.3 | Intron | NP_000848.1 | ||||
NM_001291951.1 | Intron | NP_001278880.1 | ||||
NM_001291952.1 | Intron | NP_001278881.1 | ||||
NM_001291953.1 | Intron | NP_001278882.1 | ||||
NM_001291954.1 | Intron | NP_001278883.1 | ||||
NM_001291955.1 | Intron | NP_001278884.1 | ||||
XM_011531901.1 | Intron | XP_011530203.1 | ||||
XM_017008130.1 | Intron | XP_016863619.1 | ||||
XM_017008131.1 | Intron | XP_016863620.1 | ||||
XM_017008132.1 | Intron | XP_016863621.1 | ||||
XM_017008133.1 | Intron | XP_016863622.1 |