Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTACCATGTCATGAGTTAGTCAAC[A/G]TAGGTTTGCTTTAAAATAATTATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613331 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MARCH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MARCH1 - membrane associated ring-CH-type finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166373.1 | 5091 | UTR 3 | NP_001159845.1 | |||
NM_017923.3 | 5091 | UTR 3 | NP_060393.1 | |||
XM_011532055.2 | 5091 | Intron | XP_011530357.1 | |||
XM_017008334.1 | 5091 | Intron | XP_016863823.1 | |||
XM_017008335.1 | 5091 | Intron | XP_016863824.1 | |||
XM_017008336.1 | 5091 | Intron | XP_016863825.1 |
TMA16 - translation machinery associated 16 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |