Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCTCAAACAAACCAATGAATGC[C/G]ACGACGGAGACAGCAGCGTGCCCCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604046 | |||||||||||||||||||||||
Literature Links: |
OXSR1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
OXSR1 - oxidative stress responsive 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005109.2 | 161 | Intron | NP_005100.1 | |||
XM_005265638.2 | 161 | Intron | XP_005265695.1 | |||
XM_011534331.2 | 161 | UTR 5 | XP_011532633.1 | |||
XM_017007601.1 | 161 | Intron | XP_016863090.1 | |||
XM_017007602.1 | 161 | Intron | XP_016863091.1 |