Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTTGATTTTGTTCATTAGTGGA[C/T]TGAAAACTAGGTTATGCTTTTCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611356 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GKAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GKAP1 - G kinase anchoring protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135953.1 | Intron | NP_001129425.1 | ||||
NM_025211.3 | Intron | NP_079487.2 | ||||
XM_005252241.2 | Intron | XP_005252298.1 | ||||
XM_006717293.1 | Intron | XP_006717356.1 | ||||
XM_011519058.2 | Intron | XP_011517360.1 | ||||
XM_017015171.1 | Intron | XP_016870660.1 |