Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACAAACAAACAAACAAAAAAAA[A/C]CAGAAAATGTGGGAAGTTAAGTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300131 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLS3 - plastin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136025.4 | Intron | NP_001129497.1 | ||||
NM_001172335.2 | Intron | NP_001165806.1 | ||||
NM_001282337.1 | Intron | NP_001269266.1 | ||||
NM_001282338.1 | Intron | NP_001269267.1 | ||||
NM_005032.6 | Intron | NP_005023.2 | ||||
XM_011537534.1 | Intron | XP_011535836.1 |
PLS3-AS1 - PLS3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |