Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATATAATCTTAATACATAAAAAT[A/T]AAAAACCAGAAAGAATACAATTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601244 | ||||||||||||||||||||
Literature Links: |
GUCY1A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GUCY1A2 - guanylate cyclase 1 soluble subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000855.2 | 14012 | UTR 3 | NP_000846.1 | |||
NM_001256424.1 | 14012 | UTR 3 | NP_001243353.1 |