Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612806 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR89B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR89B - G protein-coupled receptor 89B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016334.3 | Intron | NP_057418.1 | ||||
XM_005277401.3 | Intron | XP_005277458.1 | ||||
XM_005277402.4 | Intron | XP_005277459.1 | ||||
XM_005277404.3 | Intron | XP_005277461.1 | ||||
XM_006711377.3 | Intron | XP_006711440.1 | ||||
XM_006711378.2 | Intron | XP_006711441.1 | ||||
XM_011509610.2 | Intron | XP_011507912.1 | ||||
XM_011509612.2 | Intron | XP_011507914.1 | ||||
XM_011509613.2 | Intron | XP_011507915.1 | ||||
XM_017001448.1 | Intron | XP_016856937.1 | ||||
XM_017001449.1 | Intron | XP_016856938.1 |