Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGCCTTCCCTTAGACTGGCGCT[A/C]ATCTGATACTCCCTGGGCCTCCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608459 MIM: 179095 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDKL3 - cyclin dependent kinase like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113575.1 | Intron | NP_001107047.1 | ||||
NM_001300853.1 | Intron | NP_001287782.1 | ||||
NM_016508.3 | Intron | NP_057592.2 |
UBE2B - ubiquitin conjugating enzyme E2 B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003337.3 | Intron | NP_003328.1 |