Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTGTGCGCCTCCTCCGCGTGGC[C/T]GTTCCTGAATACGCTGGTGTCTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF718 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF718 - zinc finger protein 718 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039127.5 | 743 | Intron | NP_001034216.2 | |||
NM_001289930.1 | 743 | UTR 5 | NP_001276859.1 | |||
NM_001289931.1 | 743 | Intron | NP_001276860.1 | |||
XM_017007979.1 | 743 | Intron | XP_016863468.1 | |||
XM_017007980.1 | 743 | Intron | XP_016863469.1 | |||
XM_017007981.1 | 743 | Intron | XP_016863470.1 |