Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGTTTTCTTTTTTCTTTTTGTAG[A/T]AACATGTTTAACTATTTTCTCATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 125505 | ||||||||||||||||||||
Literature Links: |
DNASE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNASE1 - deoxyribonuclease I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005223.3 | Intron | NP_005214.2 | ||||
XM_005255148.3 | Intron | XP_005255205.1 | ||||
XM_006720854.3 | Intron | XP_006720917.1 | ||||
XM_011522393.2 | Intron | XP_011520695.1 | ||||
XM_017022992.1 | Intron | XP_016878481.1 | ||||
XM_017022993.1 | Intron | XP_016878482.1 | ||||
XM_017022994.1 | Intron | XP_016878483.1 | ||||
XM_017022995.1 | Intron | XP_016878484.1 | ||||
XM_017022996.1 | Intron | XP_016878485.1 | ||||
XM_017022997.1 | Intron | XP_016878486.1 | ||||
XM_017022998.1 | Intron | XP_016878487.1 | ||||
XM_017022999.1 | Intron | XP_016878488.1 | ||||
XM_017023000.1 | Intron | XP_016878489.1 | ||||
XM_017023001.1 | Intron | XP_016878490.1 | ||||
XM_017023002.1 | Intron | XP_016878491.1 | ||||
XM_017023003.1 | Intron | XP_016878492.1 | ||||
XM_017023004.1 | Intron | XP_016878493.1 | ||||
XM_017023005.1 | Intron | XP_016878494.1 | ||||
XM_017023006.1 | Intron | XP_016878495.1 | ||||
XM_017023007.1 | Intron | XP_016878496.1 |