Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCACATCCAAGCGCACCCACAA[G/T]GGGGACCCTCTGCCATCTCTTGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610503 MIM: 141251 | ||||||||||||||||||||
Literature Links: |
CDIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDIP1 - cell death-inducing p53 target 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199054.1 | 1500 | UTR 3 | NP_001185983.1 | |||
NM_001199055.1 | 1500 | UTR 3 | NP_001185984.1 | |||
NM_001199056.1 | 1500 | UTR 3 | NP_001185985.1 | |||
NM_013399.2 | 1500 | UTR 3 | NP_037531.2 |
HMOX2 - heme oxygenase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |