Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTGTCTTGTCATATTCATTTTT[C/T]CTTTTTGGTCAGTCTGATAGGTTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 314998 | ||||||||||||||||||||
Literature Links: |
ZNF81 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF81 - zinc finger protein 81 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007137.3 | Intron | NP_009068.2 | ||||
XM_005272600.3 | Intron | XP_005272657.1 | ||||
XM_011543899.2 | Intron | XP_011542201.1 | ||||
XM_011543900.2 | Intron | XP_011542202.1 | ||||
XM_017029486.1 | Intron | XP_016884975.1 | ||||
XM_017029487.1 | Intron | XP_016884976.1 |