Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCAGGTGGGCAAATTGAGCTCC[G/T]CACCTACGTGATGCCCAACGCACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600440 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENDOG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENDOG - endonuclease G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004435.2 | 1487 | Missense Mutation | CGC,CTC | R,L 245 | NP_004426.2 | |
XM_011518347.2 | 1487 | UTR 3 | XP_011516649.1 |
SPOUT1 - SPOUT domain containing methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016390.3 | 1487 | UTR 3 | NP_057474.2 | |||
XM_017014804.1 | 1487 | UTR 3 | XP_016870293.1 |