Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACCTCAGAGAACACTGATGCGCT[C/T]CGCCAGCCCCTGCATGTCGTGGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607074 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DACT3-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DACT3-AS1 - DACT3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKD2 - protein kinase D2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079880.1 | 3095 | Missense Mutation | AAG,GAG | K,E 873 | NP_001073349.1 | |
NM_001079881.1 | 3095 | Missense Mutation | AAG,GAG | K,E 873 | NP_001073350.1 | |
NM_001079882.1 | 3095 | Missense Mutation | AAG,GAG | K,E 716 | NP_001073351.1 | |
NM_016457.4 | 3095 | Missense Mutation | AAG,GAG | K,E 873 | NP_057541.2 | |
XM_005258716.2 | 3095 | Missense Mutation | AAG,GAG | K,E 716 | XP_005258773.2 |