Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCAGCGCTGCTCCAGCACCTGCA[C/T]GTGCATTCGGGCGAGCGTCCCTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605248 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCOLN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MCOLN1 - mucolipin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF358 - zinc finger protein 358 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018083.4 | 1162 | Silent Mutation | CAC,CAT | H,H 367 | NP_060553.4 | |
XM_005272460.3 | 1162 | Silent Mutation | CAC,CAT | H,H 367 | XP_005272517.1 | |
XM_011527695.1 | 1162 | Silent Mutation | CAC,CAT | H,H 367 | XP_011525997.1 | |
XM_017026296.1 | 1162 | Silent Mutation | CAC,CAT | H,H 386 | XP_016881785.1 |