Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACAGACAGAAGAGCAGCTTAGAG[C/A]CCATGACCCACCCCCTGGGAAAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD34A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD34A - ankyrin repeat domain 34A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039888.3 | 3496 | UTR 3 | NP_001034977.1 |
LIX1L - limb and CNS expressed 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR3GL - RNA polymerase III subunit G like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005277431.4 | 3496 | Intron | XP_005277488.1 | |||
XM_011510059.2 | 3496 | Intron | XP_011508361.1 |