Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGTCTCCCACCAGGGACTGGCTT[T/C]CTTCAAAGTTCCCACCGAAGGAATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 123260 | ||||||||||||||||||||
Literature Links: |
CRP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRP - C-reactive protein, pentraxin-related | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000567.2 | 601 | Missense Mutation | GAA,GGA | E,G 166 | NP_000558.2 | |
XM_011509207.2 | 601 | Missense Mutation | GAA,GGA | E,G 166 | XP_011507509.1 |