Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTAGGAGTTTTCAGAAGTATGG[C/T]TCTTGGGATAAATTTAGATTCTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601196 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRDM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRDM2 - PR domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007257.2 | 121 | Intron | NP_001007258.1 | |||
NM_001135610.1 | 121 | Intron | NP_001129082.1 | |||
NM_012231.4 | 121 | UTR 5 | NP_036363.2 | |||
NM_015866.4 | 121 | UTR 5 | NP_056950.2 | |||
XM_017002253.1 | 121 | Intron | XP_016857742.1 | |||
XM_017002254.1 | 121 | Intron | XP_016857743.1 | |||
XM_017002255.1 | 121 | Intron | XP_016857744.1 | |||
XM_017002256.1 | 121 | Intron | XP_016857745.1 | |||
XM_017002257.1 | 121 | Intron | XP_016857746.1 | |||
XM_017002258.1 | 121 | Intron | XP_016857747.1 | |||
XM_017002259.1 | 121 | Intron | XP_016857748.1 | |||
XM_017002260.1 | 121 | Intron | XP_016857749.1 | |||
XM_017002261.1 | 121 | Intron | XP_016857750.1 | |||
XM_017002262.1 | 121 | Intron | XP_016857751.1 | |||
XM_017002263.1 | 121 | Intron | XP_016857752.1 | |||
XM_017002264.1 | 121 | Intron | XP_016857753.1 |