Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCTTCTAAAGCTTTTGCATCTG[C/T]GGGAGGCTTCCTTGGGCTGCCAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605449 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSTA3 - glutathione S-transferase alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000847.4 | 527 | Missense Mutation | ACA,GCA | T,A 208 | NP_000838.3 | |
XM_006715072.3 | 527 | Missense Mutation | ACA,GCA | T,A 158 | XP_006715135.1 | |
XM_011514533.2 | 527 | Missense Mutation | ACA,GCA | T,A 115 | XP_011512835.1 | |
XM_017010794.1 | 527 | Missense Mutation | ACA,GCA | T,A 158 | XP_016866283.1 |