Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTTAATAAAAACTGTTTACTGT[C/T]ATAACCTTGGACACTTACAGGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601667 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANGPT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANGPT1 - angiopoietin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146.4 | 2775 | UTR 3 | NP_001137.2 | |||
NM_001199859.2 | 2775 | UTR 3 | NP_001186788.1 | |||
NM_001314051.1 | 2775 | UTR 3 | NP_001300980.1 |