Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGGAACCTATCATTACCCTGGA[A/G]AAGGAGGAGTGGAGGCCCAAACCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604296 MIM: 616590 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRHPR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRHPR - glyoxylate reductase/hydroxypyruvate reductase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB5 - zinc finger and BTB domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014872.2 | 1751 | Silent Mutation | TTC,TTT | F,F 517 | NP_055687.1 | |
XM_005251634.1 | 1751 | Silent Mutation | TTC,TTT | F,F 517 | XP_005251691.1 |