Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTCTTCGGCTACTCCCGAGTGGA[A/G]GTGATGCAGCAACCCTGCACCTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608168 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNH6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNH6 - potassium voltage-gated channel subfamily H member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278919.1 | 254 | Silent Mutation | GAA,GAG | E,E 58 | NP_001265848.1 | |
NM_001278920.1 | 254 | UTR 5 | NP_001265849.1 | |||
NM_030779.3 | 254 | Silent Mutation | GAA,GAG | E,E 58 | NP_110406.1 | |
NM_173092.2 | 254 | Silent Mutation | GAA,GAG | E,E 58 | NP_775115.1 | |
XM_011525308.2 | 254 | Silent Mutation | GAA,GAG | E,E 58 | XP_011523610.1 | |
XM_011525309.2 | 254 | Silent Mutation | GAA,GAG | E,E 58 | XP_011523611.1 | |
XM_011525310.2 | 254 | Silent Mutation | GAA,GAG | E,E 58 | XP_011523612.1 | |
XM_011525311.1 | 254 | Silent Mutation | GAA,GAG | E,E 58 | XP_011523613.1 | |
XM_011525312.1 | 254 | Silent Mutation | GAA,GAG | E,E 58 | XP_011523614.1 | |
XM_011525313.1 | 254 | Silent Mutation | GAA,GAG | E,E 58 | XP_011523615.1 | |
XM_017025175.1 | 254 | Silent Mutation | GAA,GAG | E,E 58 | XP_016880664.1 | |
XM_017025176.1 | 254 | Silent Mutation | GAA,GAG | E,E 58 | XP_016880665.1 | |
XM_017025177.1 | 254 | Silent Mutation | GAA,GAG | E,E 58 | XP_016880666.1 | |
XM_017025178.1 | 254 | Silent Mutation | GAA,GAG | E,E 58 | XP_016880667.1 | |
XM_017025179.1 | 254 | Silent Mutation | GAA,GAG | E,E 58 | XP_016880668.1 | |
XM_017025180.1 | 254 | UTR 5 | XP_016880669.1 |