Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCGAGGAGAGCCTTCTGGCTTGG[A/C]CTTCGGACCCCTAGAGGTTGAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNMAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PNMAL1 - paraneoplastic Ma antigen family like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001103149.1 | 1587 | Intron | NP_001096619.1 | |||
NM_018215.3 | 1587 | Missense Mutation | GCC,TCC | A,S 423 | NP_060685.2 | |
XM_011527067.2 | 1587 | Missense Mutation | GCC,TCC | A,S 423 | XP_011525369.1 |