Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAACCAAGCACAGACGACCACCA[C/T]GCTGGCAATGTCTCTGTCCACCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608740 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NFAM1 - NFAT activating protein with ITAM motif 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318323.1 | 3739 | UTR 3 | NP_001305252.1 | |||
NM_145912.6 | 3739 | UTR 3 | NP_666017.1 | |||
XM_005261374.4 | 3739 | UTR 3 | XP_005261431.1 |