Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCAACCTGAAGTTTGTGACTCT[C/T]CTGGTTGCCTTAAGTTCAGAACTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604003 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLCA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLCA2 - chloride channel accessory 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006536.5 | 216 | Silent Mutation | CTC,CTT | L,L 18 | NP_006527.1 | |
XM_011542448.2 | 216 | Silent Mutation | CTC,CTT | L,L 18 | XP_011540750.1 |