Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCCTGGCTGACCCTCCCCTGCC[C/T]GCCCACCTTCCCATGCCCCTGCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 186780 MIM: 164175 | ||||||||||||||||||||
Literature Links: |
CD247 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD247 - CD247 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000734.3 | 1286 | UTR 3 | NP_000725.1 | |||
NM_198053.2 | 1286 | UTR 3 | NP_932170.1 | |||
XM_011510144.2 | 1286 | UTR 3 | XP_011508446.1 | |||
XM_011510145.1 | 1286 | UTR 3 | XP_011508447.1 | |||
XM_017002800.1 | 1286 | UTR 3 | XP_016858289.1 | |||
XM_017002801.1 | 1286 | UTR 3 | XP_016858290.1 |
POU2F1 - POU class 2 homeobox 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |