Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGGCCCCCCATATGGCTGCATGGT[A/G]CATGGACAGTGATGAGGAACGGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605667 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COLGALT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COLGALT2 - collagen beta(1-O)galactosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303420.1 | 3332 | Intron | NP_001290349.1 | |||
NM_001303421.1 | 3332 | UTR 3 | NP_001290350.1 | |||
NM_015101.3 | 3332 | UTR 3 | NP_055916.1 | |||
XM_011509332.2 | 3332 | UTR 3 | XP_011507634.1 | |||
XM_011509334.2 | 3332 | Intron | XP_011507636.1 | |||
XM_011509335.2 | 3332 | Intron | XP_011507637.1 | |||
XM_011509336.2 | 3332 | Intron | XP_011507638.1 | |||
XM_011509337.2 | 3332 | Intron | XP_011507639.1 | |||
XM_011509338.2 | 3332 | Intron | XP_011507640.1 | |||
XM_017000750.1 | 3332 | Intron | XP_016856239.1 | |||
XM_017000751.1 | 3332 | Intron | XP_016856240.1 |
RGL1 - ral guanine nucleotide dissociation stimulator like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |