Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCTCTGCTGAGACCTATAGGGC[C/T]TCACATGCATGCAGTGCAGTCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605877 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCRL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FCRL5 - Fc receptor like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195388.1 | 4177 | UTR 3 | NP_001182317.1 | |||
NM_031281.2 | 4177 | UTR 3 | NP_112571.2 | |||
XM_011510030.1 | 4177 | Intron | XP_011508332.1 | |||
XM_011510031.1 | 4177 | Intron | XP_011508333.1 | |||
XM_011510032.1 | 4177 | Intron | XP_011508334.1 | |||
XM_011510033.1 | 4177 | Intron | XP_011508335.1 |