Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCGGGTCCCATGTCCCCTGAAGA[A/C]CCTCTCGCTTTTCCCACAGACACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615776 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CROCC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CROCC - ciliary rootlet coiled-coil, rootletin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014675.4 | Intron | NP_055490.4 | ||||
XM_006711058.3 | Intron | XP_006711121.1 | ||||
XM_011542468.1 | Intron | XP_011540770.1 | ||||
XM_011542470.2 | Intron | XP_011540772.1 | ||||
XM_017002913.1 | Intron | XP_016858402.1 | ||||
XM_017002914.1 | Intron | XP_016858403.1 | ||||
XM_017002915.1 | Intron | XP_016858404.1 |