Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTACCTTCCCACCCAGGCCCTGGT[G/T]GGAGCCCGGCCAACCCCTTTAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600244 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDCD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDCD1 - programmed cell death 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005018.2 | 1890 | UTR 3 | NP_005009.2 | |||
XM_006712573.2 | 1890 | Intron | XP_006712636.1 | |||
XM_017004293.1 | 1890 | UTR 3 | XP_016859782.1 |