Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCAACTGATTTCCTTTTCTAAG[A/G]CCACCTTCCTCTTGTGATCATTTGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603591 | |||||||||||||||||||||||
Literature Links: |
USP13 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
USP13 - ubiquitin specific peptidase 13 (isopeptidase T-3) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003940.2 | Intron | NP_003931.2 | ||||
XM_011513269.1 | Intron | XP_011511571.1 | ||||
XM_017007425.1 | Intron | XP_016862914.1 | ||||
XM_017007426.1 | Intron | XP_016862915.1 | ||||
XM_017007427.1 | Intron | XP_016862916.1 |