Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAATAAGGGAGGACTACCGTATA[T/C]ACATGTAGGGCATGACATAAAATGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604915 | |||||||||||||||||||||||
Literature Links: |
MAP3K13 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MAP3K13 - mitogen-activated protein kinase kinase kinase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242314.1 | 6578 | UTR 3 | NP_001229243.1 | |||
NM_001242317.1 | 6578 | UTR 3 | NP_001229246.1 | |||
NM_004721.4 | 6578 | UTR 3 | NP_004712.1 | |||
XM_011513310.2 | 6578 | Intron | XP_011511612.1 | |||
XM_017007456.1 | 6578 | UTR 3 | XP_016862945.1 | |||
XM_017007457.1 | 6578 | Intron | XP_016862946.1 | |||
XM_017007458.1 | 6578 | Intron | XP_016862947.1 | |||
XM_017007459.1 | 6578 | Intron | XP_016862948.1 |
TMEM41A - transmembrane protein 41A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080652.3 | 6578 | Intron | NP_542383.1 | |||
XM_017007437.1 | 6578 | Intron | XP_016862926.1 |