Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCGACCCTCCTGCGCGCCGCCTG[C/T]GCCAACAACGTGGGGCTGCTGCGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ANKRD33B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD33B - ankyrin repeat domain 33B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164440.1 | 427 | Silent Mutation | TGC,TGT | C,C 94 | NP_001157912.1 | |
XM_005248343.4 | 427 | Intron | XP_005248400.1 | |||
XM_005248344.3 | 427 | Intron | XP_005248401.1 |