Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGAGAAAATGTGTGAATTTAAT[A/C]AAAATGGGGATACCTATTCTCACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608191 | ||||||||||||||||||||
Literature Links: |
RBAK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RBAK - RB associated KRAB zinc finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204456.1 | 868 | Missense Mutation | AAA,CAA | K,Q 182 | NP_001191385.1 | |
NM_021163.3 | 868 | Missense Mutation | AAA,CAA | K,Q 182 | NP_066986.1 |
RBAK-RBAKDN - RBAK-RBAKDN readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204513.1 | 868 | Intron | NP_001191442.1 |
RBAKDN - RBAK downstream neighbor (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |