Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCCAGAAGCCTTATTTTCTGCA[C/T]GGGAAATAAGGATTAGGAAGACCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604452 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMD5 - proteasome 26S subunit, non-ATPase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270427.1 | Intron | NP_001257356.1 | ||||
NM_005047.3 | Intron | NP_005038.1 | ||||
XM_011518866.2 | Intron | XP_011517168.1 | ||||
XM_017014927.1 | Intron | XP_016870416.1 | ||||
XM_017014928.1 | Intron | XP_016870417.1 |