Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATGCAGACCTTGTAACATTGTC[C/T]ACATCACACAAGGCACCAAGGACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611392 MIM: 129010 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADO - 2-aminoethanethiol dioxygenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EGR2 - early growth response 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000399.4 | 2514 | UTR 3 | NP_000390.2 | |||
NM_001136177.2 | 2514 | UTR 3 | NP_001129649.1 | |||
NM_001136178.1 | 2514 | UTR 3 | NP_001129650.1 | |||
NM_001136179.2 | 2514 | UTR 3 | NP_001129651.1 | |||
NM_001321037.1 | 2514 | UTR 3 | NP_001307966.1 | |||
XM_011539427.1 | 2514 | UTR 3 | XP_011537729.1 |