Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACAAAGTAGAAGAAATAATACAG[A/G]ATTAAAACTGCAAAAGTAGTTAATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607669 MIM: 603674 | ||||||||||||||||||||
Literature Links: |
ARL6IP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARL6IP1 - ADP ribosylation factor like GTPase 6 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313858.1 | 2344 | UTR 3 | NP_001300787.1 | |||
NM_015161.2 | 2344 | UTR 3 | NP_055976.1 |
RPS15A - ribosomal protein S15a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |