Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAAAGCCACTGCAAATAACGCTG[C/T]GAACACAAAGCACTGGTGGCCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608124 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
XYLT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
XYLT1 - xylosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022166.3 | 5401 | UTR 3 | NP_071449.1 | |||
XM_017023539.1 | 5401 | Intron | XP_016879028.1 | |||
XM_017023540.1 | 5401 | Intron | XP_016879029.1 |