Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATGACAGCAGCCACCCCAGCCAC[C/T]TGACCCACAGCAGCACCTTCTGCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613395 MIM: 600968 | ||||||||||||||||||||
Literature Links: |
MIR138-2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR138-2 - microRNA 138-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC12A3 - solute carrier family 12 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000339.2 | 162 | Silent Mutation | CTG,TTG | L,L 45 | NP_000330.2 | |
NM_001126107.1 | 162 | Silent Mutation | CTG,TTG | L,L 45 | NP_001119579.1 | |
NM_001126108.1 | 162 | Silent Mutation | CTG,TTG | L,L 45 | NP_001119580.1 | |
XM_005256119.2 | 162 | Silent Mutation | CTG,TTG | L,L 45 | XP_005256176.1 |