Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGAGGTCCCTGGGCTGCTCCC[A/G]CAGCCCATGAGCCTCGCCTTATAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603118 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDH16 - cadherin 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204744.1 | Intron | NP_001191673.1 | ||||
NM_001204745.1 | Intron | NP_001191674.1 | ||||
NM_001204746.1 | Intron | NP_001191675.1 | ||||
NM_004062.3 | Intron | NP_004053.1 | ||||
XM_005255770.2 | Intron | XP_005255827.1 | ||||
XM_011522807.1 | Intron | XP_011521109.1 |