Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCTGAGTGGAGAGGGTAGAATG[C/T]TGGATCCCTGAATCCCACAGCTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114021 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDH3 - cadherin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317195.1 | Intron | NP_001304124.1 | ||||
NM_001317196.1 | Intron | NP_001304125.1 | ||||
NM_001793.5 | Intron | NP_001784.2 | ||||
XM_011522800.2 | Intron | XP_011521102.1 |