Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCGAGTTACCAAGTATTTTCTT[A/G]GTGAGAAATAATGTGGAGAGATGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601335 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP2K4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP2K4 - mitogen-activated protein kinase kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281435.1 | Intron | NP_001268364.1 | ||||
NM_003010.3 | Intron | NP_003001.1 | ||||
XM_005256753.2 | Intron | XP_005256810.1 | ||||
XM_005256755.2 | Intron | XP_005256812.1 | ||||
XM_011523976.2 | Intron | XP_011522278.1 | ||||
XM_011523977.2 | Intron | XP_011522279.1 |