Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCCAGCTTCCGCCGGCCCGAGG[C/T]GCCCCAGCCACTCTGGTTCATGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601463 | ||||||||||||||||||||
Literature Links: |
HAS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HAS1 - hyaluronan synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297436.1 | 1378 | Missense Mutation | ACC,GCC | T,A 489 | NP_001284365.1 | |
NM_001523.3 | 1378 | Missense Mutation | ACC,GCC | T,A 490 | NP_001514.2 | |
XM_011526884.2 | 1378 | UTR 3 | XP_011525186.1 |
SPACA6 - sperm acrosome associated 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316972.1 | 1378 | Intron | NP_001303901.1 | |||
NM_001316994.1 | 1378 | Intron | NP_001303923.1 |