Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAATTGTCTCTCACCTCAGGCTC[A/C]TTATCACCATCATGTCTGCAAAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605969 | ||||||||||||||||||||
Literature Links: |
RFPL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RFPL2 - ret finger protein like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098527.2 | 1046 | Missense Mutation | AGG,ATG | R,M 37 | NP_001091997.2 | |
NM_001159545.1 | 1046 | UTR 5 | NP_001153017.1 | |||
NM_001159546.1 | 1046 | Intron | NP_001153018.1 | |||
NM_006605.3 | 1046 | Intron | NP_006596.2 | |||
XM_011529826.1 | 1046 | Missense Mutation | AGG,ATG | R,M 37 | XP_011528128.1 | |
XM_011529827.2 | 1046 | Missense Mutation | AGG,ATG | R,M 37 | XP_011528129.1 | |
XM_011529828.1 | 1046 | Missense Mutation | AGG,ATG | R,M 37 | XP_011528130.1 | |
XM_011529830.2 | 1046 | Missense Mutation | AGG,ATG | R,M 37 | XP_011528132.1 | |
XM_011529832.1 | 1046 | Missense Mutation | AGG,ATG | R,M 37 | XP_011528134.1 | |
XM_017028535.1 | 1046 | UTR 5 | XP_016884024.1 | |||
XM_017028536.1 | 1046 | UTR 5 | XP_016884025.1 |