Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGCCCGGGGCACTCACTGAGC[A/G]CCCTCCGCTGGCTATGGCACCATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607922 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
A4GALT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
A4GALT - alpha 1,4-galactosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318038.1 | 1778 | UTR 3 | NP_001304967.1 | |||
NM_017436.5 | 1778 | UTR 3 | NP_059132.1 | |||
XM_005261644.2 | 1778 | UTR 3 | XP_005261701.1 | |||
XM_005261646.4 | 1778 | UTR 3 | XP_005261703.1 | |||
XM_005261647.2 | 1778 | UTR 3 | XP_005261704.1 | |||
XM_005261648.3 | 1778 | UTR 3 | XP_005261705.1 | |||
XM_006724265.3 | 1778 | UTR 3 | XP_006724328.1 | |||
XM_006724266.3 | 1778 | UTR 3 | XP_006724329.1 | |||
XM_011530233.2 | 1778 | UTR 3 | XP_011528535.1 | |||
XM_017028831.1 | 1778 | UTR 3 | XP_016884320.1 |