Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGAAGCCCCCACCACCAGGGGC[C/T]GGGCATGCCCAGGGCAGGTGTGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612048 MIM: 613208 | ||||||||||||||||||||
Literature Links: |
TMEM43 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM43 - transmembrane protein 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XPC - XPC complex subunit, DNA damage recognition and repair factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004628.4 | 3005 | UTR 3 | NP_004619.3 | |||
XM_011534093.2 | 3005 | Intron | XP_011532395.1 |